Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTTGAATTATGCCCTATTATAA[C/T]AATTCTGATTTTGTGGCAAGCAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603502 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFRD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFRD1 - interferon related developmental regulator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LSMEM1 - leucine rich single-pass membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134468.1 | Intron | NP_001127940.1 | ||||
NM_182597.2 | Intron | NP_872403.1 | ||||
XM_011516074.1 | Intron | XP_011514376.1 | ||||
XM_011516075.2 | Intron | XP_011514377.1 | ||||
XM_011516076.2 | Intron | XP_011514378.1 | ||||
XM_017012028.1 | Intron | XP_016867517.1 |