Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGACTTGGAAATTTTCCTCTTAA[A/T]TTTTTTAAAATTTAAAGGCAGTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142409 | ||||||||||||||||||||
Literature Links: |
HGF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HGF - hepatocyte growth factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000601.5 | 3510 | UTR 3 | NP_000592.3 | |||
NM_001010931.2 | 3510 | Intron | NP_001010931.1 | |||
NM_001010932.2 | 3510 | UTR 3 | NP_001010932.1 | |||
NM_001010933.2 | 3510 | Intron | NP_001010933.1 | |||
NM_001010934.2 | 3510 | Intron | NP_001010934.1 | |||
XM_006715956.2 | 3510 | UTR 3 | XP_006716019.1 | |||
XM_011516115.2 | 3510 | UTR 3 | XP_011514417.1 | |||
XM_017012097.1 | 3510 | Intron | XP_016867586.1 | |||
XM_017012098.1 | 3510 | Intron | XP_016867587.1 |