Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAGAATCCCCTTTTTCCCCCAA[A/C]AAAATATCTAGAGGAGCTAATAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604712 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRM2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RRM2B - ribonucleotide reductase regulatory TP53 inducible subunit M2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172477.1 | 2010 | UTR 3 | NP_001165948.1 | |||
NM_001172478.1 | 2010 | UTR 3 | NP_001165949.1 | |||
NM_015713.4 | 2010 | UTR 3 | NP_056528.2 |
Set Membership: |
HapMap |