Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTCTTGGACTGCCCCCGGGTAA[C/G]ATGCCCTAGAGATCTTTTTTTAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611145 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AARD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AARD - alanine and arginine rich domain containing protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025357.2 | Intron | NP_001020528.1 | ||||
XM_011517043.2 | Intron | XP_011515345.1 |
SLC30A8 - solute carrier family 30 member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |