Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGCTTAGTAAAAGATGAAGTTT[A/G]GTGGAGAATGAACATCCGCTGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606604 | ||||||||||||||||||||
Literature Links: |
FBXO32 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO32 - F-box protein 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242463.1 | 5748 | UTR 3 | NP_001229392.1 | |||
NM_058229.3 | 5748 | UTR 3 | NP_478136.1 | |||
NM_148177.2 | 5748 | UTR 3 | NP_680482.1 |