Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTGCCCGGCTGTGTGTCTGCCA[A/G]CCCCTTCATCCCTATCCTGGCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606449 | ||||||||||||||||||||
Literature Links: |
PTP4A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTP4A3 - protein tyrosine phosphatase type IVA, member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007079.3 | Intron | NP_009010.2 | ||||
NM_032611.2 | Intron | NP_116000.1 | ||||
XM_005250764.2 | Intron | XP_005250821.1 | ||||
XM_006716500.2 | Intron | XP_006716563.1 | ||||
XM_011516803.1 | Intron | XP_011515105.1 | ||||
XM_011516805.1 | Intron | XP_011515107.1 | ||||
XM_017012998.1 | Intron | XP_016868487.1 | ||||
XM_017012999.1 | Intron | XP_016868488.1 |