Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGAGTGTAGGAATTTTGAGAAT[C/G]TAACAACTAGATTCAAAGTACTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603775 MIM: 611351 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNE2 - cyclin E2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_057749.2 | 2057 | UTR 3 | NP_477097.1 | |||
XM_011517366.2 | 2057 | Intron | XP_011515668.1 | |||
XM_017013957.1 | 2057 | UTR 3 | XP_016869446.1 | |||
XM_017013958.1 | 2057 | UTR 3 | XP_016869447.1 | |||
XM_017013959.1 | 2057 | UTR 3 | XP_016869448.1 |
INTS8 - integrator complex subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017864.3 | 2057 | Intron | NP_060334.2 | |||
XM_017013616.1 | 2057 | Intron | XP_016869105.1 | |||
XM_017013617.1 | 2057 | Intron | XP_016869106.1 | |||
XM_017013618.1 | 2057 | UTR 3 | XP_016869107.1 | |||
XM_017013619.1 | 2057 | UTR 3 | XP_016869108.1 |