Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAAAGAATACAGTCTTCCCAAG[C/T]ATACATGTAAGGTTAGATCACATGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EFCAB1 - EF-hand calcium binding domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142857.1 | Intron | NP_001136329.1 | ||||
NM_024593.3 | Intron | NP_078869.1 | ||||
XM_005251303.1 | Intron | XP_005251360.1 | ||||
XM_011517589.1 | Intron | XP_011515891.1 | ||||
XM_011517590.2 | Intron | XP_011515892.1 | ||||
XM_011517591.2 | Intron | XP_011515893.1 | ||||
XM_017013826.1 | Intron | XP_016869315.1 | ||||
XM_017013827.1 | Intron | XP_016869316.1 | ||||
XM_017013828.1 | Intron | XP_016869317.1 |