Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGCAGGTATTGGTGTGAACCTGG[A/T]GGTATTTCTTTGGTTTTCCAGTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
YTHDF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
YTHDF3 - YTH N6-methyladenosine RNA binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277813.1 | Intron | NP_001264742.1 | ||||
NM_001277814.1 | Intron | NP_001264743.1 | ||||
NM_001277815.1 | Intron | NP_001264744.1 | ||||
NM_001277816.1 | Intron | NP_001264745.1 | ||||
NM_001277817.1 | Intron | NP_001264746.1 | ||||
NM_001277818.1 | Intron | NP_001264747.1 | ||||
NM_152758.5 | Intron | NP_689971.4 | ||||
XM_011517509.2 | Intron | XP_011515811.1 | ||||
XM_011517510.2 | Intron | XP_011515812.1 |
YTHDF3-AS1 - YTHDF3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |