Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AACTGATGAAATTGAAGGACTACTA[A/C]ATAAATGGAGATATATTTCATGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604141 MIM: 611654 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFGEF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARFGEF1 - ADP ribosylation factor guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006421.4 | Intron | NP_006412.2 | ||||
XM_005251134.4 | Intron | XP_005251191.1 | ||||
XM_005251135.3 | Intron | XP_005251192.1 | ||||
XM_005251136.3 | Intron | XP_005251193.1 | ||||
XM_011517442.2 | Intron | XP_011515744.1 |