Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTGCCATCTGCTTTGAAAACGT[C/T]TCATTCATTTTAGGGCCGAAGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612139 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PREX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PREX2 - phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024870.3 | Intron | NP_079146.2 | ||||
NM_025170.5 | Intron | NP_079446.3 | ||||
XM_011517612.2 | Intron | XP_011515914.1 |