Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603026 | ||||||||||||||||||||
Literature Links: |
PLAG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLAG1 - PLAG1 zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114634.1 | 4584 | UTR 3 | NP_001108106.1 | |||
NM_001114635.1 | 4584 | UTR 3 | NP_001108107.1 | |||
NM_002655.2 | 4584 | UTR 3 | NP_002646.2 | |||
XM_011517544.2 | 4584 | UTR 3 | XP_011515846.1 | |||
XM_017013576.1 | 4584 | UTR 3 | XP_016869065.1 | |||
XM_017013577.1 | 4584 | UTR 3 | XP_016869066.1 |