Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAATGAGCTTTCCCACTTTTTAAG[C/G]CTTCCTGATTTCCCGTTTGAGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611328 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJB5 - DnaJ heat shock protein family (Hsp40) member B5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135004.2 | Intron | NP_001128476.2 | ||||
NM_001135005.2 | Intron | NP_001128477.1 | ||||
NM_012266.5 | Intron | NP_036398.3 | ||||
XM_005251428.3 | Intron | XP_005251485.1 | ||||
XM_006716751.2 | Intron | XP_006716814.1 | ||||
XM_011517842.2 | Intron | XP_011516144.1 |
DNAJB5-AS1 - DNAJB5 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |