Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGCTTTTTTGTTACCCATCCTC[A/G]TATGTCCCAGCTAGAAAGAATCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605723 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDCD1LG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDCD1LG2 - programmed cell death 1 ligand 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025239.3 | 87 | UTR 5 | NP_079515.2 | |||
XM_005251600.3 | 87 | UTR 5 | XP_005251657.1 |