Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGAGACCCATCCGTTATTTTCC[A/C]TCTGAAGCTGGAAAGAGTTAAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604785 MIM: 603722 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTNNAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTNNAL1 - catenin alpha like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286974.1 | 2336 | UTR 3 | NP_001273903.1 | |||
NM_003798.3 | 2336 | Missense Mutation | AGG,ATG | R,M 716 | NP_003789.1 | |
XM_005252291.3 | 2336 | Missense Mutation | AGG,ATG | R,M 714 | XP_005252348.1 | |
XM_017015250.1 | 2336 | Missense Mutation | AGG,ATG | R,M 539 | XP_016870739.1 | |
XM_017015251.1 | 2336 | Missense Mutation | AGG,ATG | R,M 537 | XP_016870740.1 |
FAM206A - family with sequence similarity 206 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017832.3 | 2336 | Intron | NP_060302.1 | |||
XM_011518816.2 | 2336 | Intron | XP_011517118.1 |
IKBKAP - inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |