Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCTGGTGGGCGGCCGCGAGGCT[C/T]GGGAGAGGCGAACCGGAGCGCGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602874 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGCG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UGCG - UDP-glucose ceramide glucosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003358.2 | 156 | UTR 5 | NP_003349.1 | |||
XM_005252186.4 | 156 | UTR 5 | XP_005252243.1 | |||
XM_017015107.1 | 156 | Intron | XP_016870596.1 |