Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTGAGCTAGTGCCATTGGGGGT[G/T]GGGGGTAAAAGCTATTCTTATTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603088 | ||||||||||||||||||||
Literature Links: |
SLC31A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC31A2 - solute carrier family 31 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001860.2 | 1695 | Intron | NP_001851.1 | |||
XM_011518216.1 | 1695 | Intron | XP_011516518.1 | |||
XM_017014267.1 | 1695 | UTR 5 | XP_016869756.1 |