Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGATACATGAGGGGACAAAAT[G/T]TGCCCCATTCACCCTCATATTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603088 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKBP15 - FK506 binding protein 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015258.1 | Intron | NP_056073.1 | ||||
XM_006717018.2 | Intron | XP_006717081.1 | ||||
XM_006717019.1 | Intron | XP_006717082.1 | ||||
XM_017014552.1 | Intron | XP_016870041.1 |
SLC31A2 - solute carrier family 31 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |