Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACGTGGCCTATTCTGCAGAGCT[C/T]CTTCATCTCCAGGAACAGAAAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC107987125 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107987125 - uncharacterized LOC107987125 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTLL11 - tubulin tyrosine ligase like 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139442.1 | 7648 | Intron | NP_001132914.1 | |||
NM_194252.2 | 7648 | Intron | NP_919228.2 | |||
XM_005251728.2 | 7648 | UTR 3 | XP_005251785.1 | |||
XM_005251729.4 | 7648 | UTR 3 | XP_005251786.1 | |||
XM_006716966.3 | 7648 | Intron | XP_006717029.1 | |||
XM_017014309.1 | 7648 | Intron | XP_016869798.1 |