Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCTATACCTGTCACTGGTATCT[A/G]CAAACACATCCACCAAGAATTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612693 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
URM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
URM1 - ubiquitin related modifier 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135947.2 | Intron | NP_001129419.1 | ||||
NM_001265582.1 | Intron | NP_001252511.1 | ||||
NM_030914.3 | Intron | NP_112176.1 |