Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGAGATAGTGGCTGAAATGGAT[G/T]TTCCTCTCCACTGCCGCACAGAATT
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 608360 | ||||||||||||||||||||||||||
Literature Links: |
LRRC8A PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
LRRC8A - leucine rich repeat containing 8 family member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHYHD1 - phytanoyl-CoA dioxygenase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100876.1 | 1066 | Missense Mutation | GTT,TTT | V,F 45 | NP_001094346.1 | |
NM_001100877.1 | 1066 | Missense Mutation | GTT,TTT | V,F 45 | NP_001094347.1 | |
NM_174933.3 | 1066 | Missense Mutation | GTT,TTT | V,F 45 | NP_777593.2 |
Set Membership: |
HapMap |