Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTAGAGAGCAAACGCAGGCAGAA[A/G]TAAAAACACCAGATAAAAAGTGAGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615587 MIM: 609288 | |||||||||||||||||||||||
Literature Links: |
NUP188 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NUP188 - nucleoporin 188 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015354.2 | 2042 | Intron | NP_056169.1 |
SH3GLB2 - SH3 domain containing GRB2 like endophilin B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287045.1 | 2042 | UTR 3 | NP_001273974.1 | |||
NM_001287046.1 | 2042 | UTR 3 | NP_001273975.1 | |||
NM_020145.3 | 2042 | UTR 3 | NP_064530.1 | |||
XM_005252098.2 | 2042 | UTR 3 | XP_005252155.1 | |||
XM_005252100.2 | 2042 | UTR 3 | XP_005252157.1 | |||
XM_005252101.3 | 2042 | UTR 3 | XP_005252158.1 | |||
XM_006717188.2 | 2042 | UTR 3 | XP_006717251.1 | |||
XM_006717189.2 | 2042 | UTR 3 | XP_006717252.1 | |||
XM_006717190.1 | 2042 | UTR 3 | XP_006717253.1 | |||
XM_006717191.1 | 2042 | UTR 3 | XP_006717254.1 |