Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCTCTCAGCTCCCCTACCTTGA[C/T]GCCCCCAGCACTTCCCAGGCTGCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600184 MIM: 614516 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRAT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CRAT - carnitine O-acetyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000755.3 | Intron | NP_000746.2 | ||||
NM_001257363.1 | Intron | NP_001244292.1 | ||||
XM_005251708.3 | Intron | XP_005251765.1 | ||||
XM_017014274.1 | Intron | XP_016869763.1 | ||||
XM_017014275.1 | Intron | XP_016869764.1 |
DOLPP1 - dolichyldiphosphatase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |