Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600184 MIM: 614516 | ||||||||||||||||||||
Literature Links: |
CRAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRAT - carnitine O-acetyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000755.3 | 1931 | Missense Mutation | GAT,GGT | D,G 517 | NP_000746.2 | |
NM_001257363.1 | 1931 | Missense Mutation | GAT,GGT | D,G 496 | NP_001244292.1 | |
XM_005251708.3 | 1931 | Missense Mutation | GAT,GGT | D,G 496 | XP_005251765.1 | |
XM_017014274.1 | 1931 | Missense Mutation | GAT,GGT | D,G 518 | XP_016869763.1 | |
XM_017014275.1 | 1931 | Missense Mutation | GAT,GGT | D,G 518 | XP_016869764.1 |
DOLPP1 - dolichyldiphosphatase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |