Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCGTTCTGGAAAGAGGTGATACA[C/T]TATTTCCCTTCTAAGAGACAGCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF510 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF510 - zinc finger protein 510 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001314059.1 | 4642 | UTR 3 | NP_001300988.1 | |||
NM_001314060.1 | 4642 | UTR 3 | NP_001300989.1 | |||
NM_014930.2 | 4642 | UTR 3 | NP_055745.1 | |||
XM_005251808.2 | 4642 | UTR 3 | XP_005251865.1 | |||
XM_011518393.2 | 4642 | UTR 3 | XP_011516695.1 | |||
XM_017014483.1 | 4642 | UTR 3 | XP_016869972.1 |