Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCTTCCAAGTCTAGAATTTGCA[A/G]ACCAGTCTATGTCCTCTGTGACTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C10orf107 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C10orf107 - chromosome 10 open reading frame 107 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173554.2 | Intron | NP_775825.1 | ||||
XM_005269598.3 | Intron | XP_005269655.1 | ||||
XM_005269599.4 | Intron | XP_005269656.1 | ||||
XM_005269600.4 | Intron | XP_005269657.1 | ||||
XM_006717690.3 | Intron | XP_006717753.1 | ||||
XM_006717692.3 | Intron | XP_006717755.1 |