Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCCTGGGGGTGGGGCTGTAGGG[A/G]AAAGTGCTAAAGCCGCTGAGGTAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613427 MIM: 614215 | ||||||||||||||||||||
Literature Links: |
ANAPC16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANAPC16 - anaphase promoting complex subunit 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242546.1 | 119 | UTR 5 | NP_001229475.1 | |||
NM_001242547.1 | 119 | Intron | NP_001229476.1 | |||
NM_001242548.1 | 119 | Intron | NP_001229477.1 | |||
NM_173473.3 | 119 | UTR 5 | NP_775744.1 |
ASCC1 - activating signal cointegrator 1 complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198798.2 | 119 | Intron | NP_001185727.1 | |||
NM_001198799.2 | 119 | Intron | NP_001185728.1 | |||
NM_001198800.2 | 119 | Intron | NP_001185729.1 |