Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATCTGTGCAGAATGCCCTCTTC[C/T]GGCCATATTCTTTGACCTGACTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602809 | ||||||||||||||||||||
Literature Links: |
KIF5B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF5B - kinesin family member 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004521.2 | 2813 | Missense Mutation | AGA,GGA | R,G 913 | NP_004512.1 | |
XM_017016224.1 | 2813 | Missense Mutation | AGA,GGA | R,G 819 | XP_016871713.1 |