Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATTGACTAATCTTTTCTTAAAG[C/G]CTTCAGTGCACTTGCACGTGACTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608734 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC39A12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC39A12 - solute carrier family 39 member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145195.1 | Intron | NP_001138667.1 | ||||
NM_001282733.1 | Intron | NP_001269662.1 | ||||
NM_001282734.1 | Intron | NP_001269663.1 | ||||
NM_152725.3 | Intron | NP_689938.2 |