Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATTTTTATGCATGCTTTAAACA[A/G]ACAGTATTTTTTTTAAATGAGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603050 MIM: 607429 | ||||||||||||||||||||
Literature Links: |
ABI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABI1 - abl interactor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012750.2 | 2025 | UTR 3 | NP_001012768.1 | |||
NM_001012751.2 | 2025 | UTR 3 | NP_001012769.1 | |||
NM_001012752.2 | 2025 | UTR 3 | NP_001012770.1 | |||
NM_001178116.1 | 2025 | UTR 3 | NP_001171587.1 | |||
NM_001178119.1 | 2025 | UTR 3 | NP_001171590.1 | |||
NM_001178120.1 | 2025 | UTR 3 | NP_001171591.1 | |||
NM_001178121.1 | 2025 | UTR 3 | NP_001171592.1 | |||
NM_001178122.1 | 2025 | UTR 3 | NP_001171593.1 | |||
NM_001178123.1 | 2025 | UTR 3 | NP_001171594.1 | |||
NM_001178124.1 | 2025 | UTR 3 | NP_001171595.1 | |||
NM_001178125.1 | 2025 | UTR 3 | NP_001171596.1 | |||
NM_005470.3 | 2025 | UTR 3 | NP_005461.2 | |||
XM_005252332.3 | 2025 | UTR 3 | XP_005252389.1 | |||
XM_005252333.3 | 2025 | UTR 3 | XP_005252390.1 | |||
XM_006717359.2 | 2025 | UTR 3 | XP_006717422.1 | |||
XM_006717360.2 | 2025 | UTR 3 | XP_006717423.1 | |||
XM_006717361.2 | 2025 | UTR 3 | XP_006717424.1 | |||
XM_011519289.2 | 2025 | Intron | XP_011517591.1 | |||
XM_017015458.1 | 2025 | UTR 3 | XP_016870947.1 | |||
XM_017015459.1 | 2025 | Intron | XP_016870948.1 |
PDSS1 - prenyl (decaprenyl) diphosphate synthase, subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321978.1 | 2025 | Intron | NP_001308907.1 | |||
NM_001321979.1 | 2025 | Intron | NP_001308908.1 | |||
NM_014317.4 | 2025 | Intron | NP_055132.2 |