Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATACCTGGGAGACACTTTAGATT[C/T]CTCTAGAAGTTTCTTGAATTCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TCERG1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCERG1L - transcription elongation regulator 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174937.3 | 1671 | Missense Mutation | AAA,GAA | K,E 529 | NP_777597.2 | |
XM_011539577.1 | 1671 | Missense Mutation | AAA,GAA | K,E 542 | XP_011537879.1 | |
XM_011539578.2 | 1671 | Intron | XP_011537880.1 |
TCERG1L-AS1 - TCERG1L antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |