Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGTGAGGATGGGAAATGTCAGA[C/G]CACGTGACAAATTTTGAAATTTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC82 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC82 - coiled-coil domain containing 82 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318736.1 | 3080 | Intron | NP_001305665.1 | |||
NM_001318737.1 | 3080 | Intron | NP_001305666.1 | |||
NM_024725.3 | 3080 | Intron | NP_079001.2 | |||
XM_005274292.3 | 3080 | Intron | XP_005274349.1 | |||
XM_006718911.3 | 3080 | Intron | XP_006718974.1 | |||
XM_011542984.2 | 3080 | Intron | XP_011541286.1 | |||
XM_011542987.2 | 3080 | Intron | XP_011541289.1 | |||
XM_011542988.2 | 3080 | Intron | XP_011541290.1 | |||
XM_017018305.1 | 3080 | Intron | XP_016873794.1 | |||
XM_017018306.1 | 3080 | Intron | XP_016873795.1 | |||
XM_017018307.1 | 3080 | Intron | XP_016873796.1 | |||
XM_017018308.1 | 3080 | Intron | XP_016873797.1 | |||
XM_017018309.1 | 3080 | UTR 3 | XP_016873798.1 | |||
XM_017018310.1 | 3080 | UTR 3 | XP_016873799.1 | |||
XM_017018311.1 | 3080 | Intron | XP_016873800.1 | |||
XM_017018312.1 | 3080 | Intron | XP_016873801.1 |