Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATGGAGGGTCATGATCTCCCCG[G/T]GTGGCAGCTGTGCATTTGATCTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610603 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACCSL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACCSL - 1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031854.2 | Intron | NP_001027025.2 | ||||
XM_017017714.1 | Intron | XP_016873203.1 | ||||
XM_017017715.1 | Intron | XP_016873204.1 |
ALKBH3 - alkB homolog 3, alpha-ketoglutaratedependent dioxygenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ALKBH3-AS1 - ALKBH3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |