Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTGCTCCTGTGGGTCACTTGAG[G/T]CAGGCTCCACCTTCCCCAGGAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102772 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMPD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMPD3 - adenosine monophosphate deaminase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000480.2 | 248 | Intron | NP_000471.1 | |||
NM_001025389.1 | 248 | UTR 5 | NP_001020560.1 | |||
NM_001025390.1 | 248 | Intron | NP_001020561.1 | |||
NM_001172430.1 | 248 | Intron | NP_001165901.1 | |||
NM_001172431.1 | 248 | Intron | NP_001165902.1 |