Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGAGGGCAGCGTCCAGAACATC[C/T]AAGTATGGGGGACCAGGAAAAGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147470 MIM: 610146 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGF2 - insulin like growth factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000612.5 | Intron | NP_000603.1 | ||||
NM_001007139.5 | Intron | NP_001007140.2 | ||||
NM_001127598.2 | Intron | NP_001121070.1 | ||||
NM_001291861.2 | Intron | NP_001278790.1 | ||||
NM_001291862.2 | Intron | NP_001278791.1 |
IGF2-AS - IGF2 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
INS-IGF2 - INS-IGF2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042376.2 | Intron | NP_001035835.1 |
MIR483 - microRNA 483 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |