Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTGGGTGTTTAGCTTTCTAATC[C/G]CATTCTTCGGTGAATTTACATTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602601 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OLR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OLR1 - oxidized low density lipoprotein receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172632.1 | Intron | NP_001166103.1 | ||||
NM_001172633.1 | Intron | NP_001166104.1 | ||||
NM_002543.3 | Intron | NP_002534.1 |
TMEM52B - transmembrane protein 52B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079815.1 | Intron | NP_001073283.1 | ||||
NM_153022.3 | Intron | NP_694567.1 | ||||
XM_006719029.2 | Intron | XP_006719092.1 |