Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGTGCGATTATCTTTGGTAATT[A/T]AAAAAAAAAAAAAAAAAAAGGCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138330 | ||||||||||||||||||||
Literature Links: |
MGST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MGST1 - microsomal glutathione S-transferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260511.1 | Intron | NP_001247440.1 | ||||
NM_001260512.1 | Intron | NP_001247441.1 | ||||
NM_001267598.1 | Intron | NP_001254527.1 | ||||
NM_020300.4 | Intron | NP_064696.1 | ||||
NM_145764.2 | Intron | NP_665707.1 | ||||
NM_145791.2 | Intron | NP_665734.1 | ||||
NM_145792.2 | Intron | NP_665735.1 | ||||
XM_006719079.1 | Intron | XP_006719142.1 | ||||
XM_011520673.1 | Intron | XP_011518975.1 | ||||
XM_011520674.2 | Intron | XP_011518976.1 |