Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCTTGAGTAGCTGGGACTACAG[C/G]CTTGGGGTTTCACCATGTTGGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607240 MIM: 614093 | ||||||||||||||||||||
Literature Links: |
KMT5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KMT5A - lysine methyltransferase 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RILPL2 - Rab interacting lysosomal protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145058.2 | Intron | NP_659495.1 | ||||
XM_011538012.2 | Intron | XP_011536314.1 | ||||
XM_017018959.1 | Intron | XP_016874448.1 |