Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACTTCCCTTTTAGAGGCGGAGC[G/T]TGCAGTGAGCCGAGATTGTGCCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608090 | ||||||||||||||||||||
Literature Links: |
MLXIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MLXIP - MLX interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014938.5 | Intron | NP_055753.3 | ||||
XM_006719290.3 | Intron | XP_006719353.1 | ||||
XM_006719291.3 | Intron | XP_006719354.1 | ||||
XM_006719292.3 | Intron | XP_006719355.1 | ||||
XM_006719293.3 | Intron | XP_006719356.1 | ||||
XM_006719294.3 | Intron | XP_006719357.1 |