Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCAGCGCACGGCCCTCACCCCG[C/T]ATCGCGACCCCGCGCCCCGCCGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615627 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRI3BP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRI3BP - BRI3 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080626.5 | 102 | UTR 5 | NP_542193.3 | |||
XM_011537940.2 | 102 | UTR 5 | XP_011536242.1 |
DHX37 - DEAH-box helicase 37 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |