Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGACTTTGATCCTAGGCTCTGC[A/G]CTGTGGGTGGTGTTATCTCAACTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608090 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLXIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MLXIP - MLX interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014938.5 | Intron | NP_055753.3 | ||||
XM_006719290.3 | Intron | XP_006719353.1 | ||||
XM_006719291.3 | Intron | XP_006719354.1 | ||||
XM_006719292.3 | Intron | XP_006719355.1 | ||||
XM_006719293.3 | Intron | XP_006719356.1 | ||||
XM_006719294.3 | Intron | XP_006719357.1 |