Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCATGGCAAATTCCATGGCACC[G/T]TCAAGGCTGAGAACGGGAAGCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138400 MIM: 610495 MIM: 615638 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAPDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAPDH - glyceraldehyde-3-phosphate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256799.2 | 458 | Missense Mutation | GTC,TTC | V,F 18 | NP_001243728.1 | |
NM_001289745.1 | 458 | Missense Mutation | NP_001276674.1 | |||
NM_001289746.1 | 458 | Intron | NP_001276675.1 | |||
NM_002046.5 | 458 | Missense Mutation | NP_002037.2 |
IFFO1 - intermediate filament family orphan 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCAPD2 - non-SMC condensin I complex subunit D2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |