Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCACCGTCAAGGCTGAGAACGG[G/T]AAGCTTGTCATCAATGGAAATCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138400 MIM: 610495 MIM: 615638 | ||||||||||||||||||||
Literature Links: |
GAPDH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAPDH - glyceraldehyde-3-phosphate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256799.2 | 475 | Silent Mutation | GGG,GGT | G,G 23 | NP_001243728.1 | |
NM_001289745.1 | 475 | Silent Mutation | NP_001276674.1 | |||
NM_001289746.1 | 475 | Intron | NP_001276675.1 | |||
NM_002046.5 | 475 | Silent Mutation | NP_002037.2 |
IFFO1 - intermediate filament family orphan 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCAPD2 - non-SMC condensin I complex subunit D2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |