Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCAGCCCCAGCGTGCACAGCTC[A/G]GTGTCCAGAGCAATGGGTGCTCCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186940 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD4 - CD4 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000616.4 | Intron | NP_000607.1 | ||||
NM_001195014.2 | Intron | NP_001181943.1 | ||||
NM_001195015.2 | Intron | NP_001181944.1 | ||||
NM_001195016.2 | Intron | NP_001181945.1 | ||||
NM_001195017.2 | Intron | NP_001181946.1 | ||||
XM_017020228.1 | Intron | XP_016875717.1 |