Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATAGTCCACTATTTTCACGCTAA[C/T]GGTGCTTCAATATTCTTCATCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601184 | ||||||||||||||||||||
Literature Links: |
DNAJC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC3 - DnaJ heat shock protein family (Hsp40) member C3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006260.4 | Intron | NP_006251.1 | ||||
XM_011521104.2 | Intron | XP_011519406.1 | ||||
XM_011521105.1 | Intron | XP_011519407.1 | ||||
XM_017020674.1 | Intron | XP_016876163.1 | ||||
XM_017020675.1 | Intron | XP_016876164.1 |