Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCCCGTGCGCCCTCTTCGATCC[G/T]GGGGTGCTCCGGTCCCTCCTCCCCG
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 605123 | |||||||||||||||||||||||||||||
Literature Links: |
SPINT1 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SPINT1 - serine peptidase inhibitor, Kunitz type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032367.1 | Intron | NP_001027539.1 | ||||
NM_003710.3 | Intron | NP_003701.1 | ||||
NM_181642.2 | Intron | NP_857593.1 | ||||
XM_006720657.1 | Intron | XP_006720720.1 | ||||
XM_011521957.1 | Intron | XP_011520259.1 | ||||
XM_011521958.1 | Intron | XP_011520260.1 |
Set Membership: |
HapMap |