Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCAGTAATAAATATCCGGGTCT[C/G]TAATAAGAGTTAGGTATTAGAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601272 | ||||||||||||||||||||
Literature Links: |
FAM96A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM96A - family with sequence similarity 96 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX1 - sorting nexin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242933.1 | Intron | NP_001229862.1 | ||||
NM_003099.4 | Intron | NP_003090.2 | ||||
NM_148955.3 | Intron | NP_683758.1 | ||||
XM_005254611.2 | Intron | XP_005254668.1 |