Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTCAAACATAGACAAAGATAGG[G/T]AGATTAGGATAATAAATCCCTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603617 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC24A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC24A1 - solute carrier family 24 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001254740.1 | Intron | NP_001241669.1 | ||||
NM_001301031.1 | Intron | NP_001287960.1 | ||||
NM_001301032.1 | Intron | NP_001287961.1 | ||||
NM_001301033.1 | Intron | NP_001287962.1 | ||||
NM_004727.2 | Intron | NP_004718.1 | ||||
XM_005254778.3 | Intron | XP_005254835.1 | ||||
XM_005254781.3 | Intron | XP_005254838.1 | ||||
XM_006720768.3 | Intron | XP_006720831.1 | ||||
XM_011522219.1 | Intron | XP_011520521.1 | ||||
XM_011522220.2 | Intron | XP_011520522.1 | ||||
XM_011522221.2 | Intron | XP_011520523.1 | ||||
XM_011522222.2 | Intron | XP_011520524.1 | ||||
XM_011522226.2 | Intron | XP_011520528.1 | ||||
XM_017022724.1 | Intron | XP_016878213.1 | ||||
XM_017022725.1 | Intron | XP_016878214.1 |
VWA9 - von Willebrand factor A domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |