Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAACCAAACCAAAACAAAAAAC[A/C]AAAAACAAAAAAAAACCCTTAGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600382 MIM: 603617 | ||||||||||||||||||||
Literature Links: |
DENND4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DENND4A - DENN domain containing 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144823.2 | 5829 | Intron | NP_001138295.1 | |||
NM_001320835.1 | 5829 | Intron | NP_001307764.1 | |||
NM_005848.4 | 5829 | Intron | NP_005839.3 | |||
XM_005254120.4 | 5829 | Intron | XP_005254177.1 | |||
XM_005254121.3 | 5829 | Intron | XP_005254178.1 | |||
XM_005254122.4 | 5829 | Intron | XP_005254179.1 | |||
XM_006720371.3 | 5829 | Intron | XP_006720434.1 | |||
XM_011521156.2 | 5829 | Intron | XP_011519458.1 | |||
XM_011521157.2 | 5829 | Intron | XP_011519459.1 | |||
XM_017021862.1 | 5829 | Intron | XP_016877351.1 | |||
XM_017021863.1 | 5829 | Intron | XP_016877352.1 | |||
XM_017021864.1 | 5829 | UTR 3 | XP_016877353.1 | |||
XM_017021865.1 | 5829 | Intron | XP_016877354.1 |
SLC24A1 - solute carrier family 24 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |