Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCAGCAGTGTTTCCTCTTCATT[A/T]AAAGTATTGGCAATAGCATCATTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614183 MIM: 610716 | ||||||||||||||||||||
Literature Links: |
DIS3L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DIS3L - DIS3 like exosome 3'-5' exoribonuclease | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIPIN - TIMELESS interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289986.1 | 1085 | Intron | NP_001276915.1 | |||
NM_017858.2 | 1085 | Intron | NP_060328.2 | |||
XM_005254521.2 | 1085 | Missense Mutation | TTA,TTT | L,F 272 | XP_005254578.1 | |
XM_005254522.4 | 1085 | Missense Mutation | TTA,TTT | L,F 272 | XP_005254579.1 | |
XM_006720594.3 | 1085 | Missense Mutation | TTA,TTT | L,F 272 | XP_006720657.1 | |
XM_011521743.1 | 1085 | Missense Mutation | TTA,TTT | L,F 272 | XP_011520045.1 | |
XM_017022390.1 | 1085 | Missense Mutation | TTA,TTT | L,F 272 | XP_016877879.1 | |
XM_017022391.1 | 1085 | Intron | XP_016877880.1 |